Recommended Products
form
solid
reaction suitability
reaction type: C-C Bond Formation
reagent type: catalyst
reaction type: C-H Activation
mp
84-85 °C
storage temp.
−20°C
InChI
1S/C10H13NOS/c1-9(12)11-7-8-13-10-5-3-2-4-6-10/h2-6H,7-8H2,1H3,(H,11,12)
InChI key
RSOVUTMFSIKQHH-UHFFFAOYSA-N
Related Categories
Application
N-(2-(Phenylthio)ethyl)acetamide is a C-H activation ligand developed by the Yu Group for the Pd-catalyzed olefination/lactonization of C(sp3)-H bonds of free carboxylic acids.
related product
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Journal of the American Chemical Society, 140(32), 10363-10367 (2018-07-22)
An acetyl-protected aminoethyl phenyl thioether has been developed to promote C(sp3)-H activation. Significant ligand enhancement is demonstrated by the realization of the first Pd(II)-catalyzed olefination of C(sp3)-H bonds of free carboxylic acids without using an auxiliary. Subsequent lactonization of the
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service