Skip to Content
Merck
All Photos(1)

Key Documents

900922

Sigma-Aldrich

2,6-Bis(9H-carbazol-9-yl)pyridine

Synonym(s):

PYD-2Cz

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H19N3
CAS Number:
Molecular Weight:
409.48
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.23

Assay

≥98%

form

solid

Looking for similar products? Visit Product Comparison Guide

Application

A highly efficient host material for blue phosphorescent OLEDs.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Excimer-based white phosphorescent organic light emitting diodes with nearly 100% internal quantum efficiency.
Williams EL, et al.
Advanced Materials, 19(2), 197-202 (2007)
Chao Tang et al.
Chemical communications (Cambridge, England), 51(9), 1650-1653 (2014-12-17)
By using various fluoro-substituted pyridines and carbazole as starting materials, we have synthesized a series of carbazole-pyridine hybrid compounds through an efficient catalyst free C-N coupling reaction with high yields of 85-95%. The bi-, tri- and tetra-carbazole substituted pyridine derivatives
Tetradentate Pt(II) complexes with 6-membered chelate rings: A new route for stable and efficient blue organic light Emitting Diodes.
Fleetham TB, et al.
Chemistry of Materials, 28(10), 3276-3282 (2016)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service