Skip to Content
Merck
All Photos(1)

Key Documents

900510

Sigma-Aldrich

Vinyl N,N-diisopropylcarbamate

Synonym(s):

Vinyl carbamate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H17NO2
CAS Number:
Molecular Weight:
171.24
UNSPSC Code:
12352101
NACRES:
NA.22

form

liquid

refractive index

n/D 1.4397

density

0.92 g/mL

storage temp.

2-8°C

Application

Alkyne-containing molecules can be prepared from boronic ester building blocks. With high enantiospecificity, this vinyl carbamate can be used to alkynylate enantioenriched secondary and tertiary boronic esters in good yields, accomplished by a Zweifel-type olefination followed by a 1,2-elimination.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

108.0 °F

Flash Point(C)

42.22 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service