Skip to Content
Merck
All Photos(2)

Key Documents

900292

Sigma-Aldrich

Poly(caprolactone-co-glycolide)

caprolactone:glycolide 90:10, viscosity 0.75 dL/g 

Synonym(s):

PCL-PGA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(C6H10O2)m(C4H4O4)nCH3
UNSPSC Code:
12162002
NACRES:
NA.23

form

chips or granules

feed ratio

caprolactone:glycolide 90:10

viscosity

0.75 dL/g

transition temp

flash point >230 °F
Tm 46-51 °C

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Innovations in polymer technology have had a significant impact on the advancement of novel drug delivery systems.

Questions

  1. What is the degradation time of this product?

    1 answer
    1. The degradation time for this material is not determined. However, it may be helpful to review the following publications on the degradation properties of PLA-PCL:
      https://www.scientific.net/MSF.636-637.825
      https://www.ncbi.nlm.nih.gov/pmc/articles/PMC8400216/

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service