860484
4-Nitrophenyl N-acetyl-β-D-glucosaminide
99%
Synonym(s):
4-Nitrophenyl-2-acetamido-2-deoxy-β-D-glucopyranoside, 4-nitrophenyl-N-acetyl-β-D-glucosaminide, p-Nitrophenyl 2-acetamido-2-deoxy-β-D-glucopyranoside
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C14H18N2O8
CAS Number:
Molecular Weight:
342.30
Beilstein:
96193
EC Number:
MDL number:
UNSPSC Code:
12352103
Assay
99%
optical activity
[α]22/D −16.0°, c = 0.5 in H2O
mp
210-212 °C (lit.)
SMILES string
CC(=O)N[C@@H]1[C@@H](O)[C@H](O)[C@@H](CO)O[C@H]1Oc2ccc(cc2)[N+]([O-])=O
replaced by
Product No.
Description
Pricing
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service