Skip to Content
Merck
All Photos(1)

Key Documents

79234

Sigma-Aldrich

(±)-α-(Trifluoromethyl)benzyl alcohol

≥98.0% (GC)

Synonym(s):

(±)-1-Phenyl-2,2,2-trifluoroethanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3CH(OH)C6H5
CAS Number:
Molecular Weight:
176.14
Beilstein:
3197623
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.0% (GC)

form

liquid

refractive index

n20/D 1.462

bp

57-59 °C/2 mmHg (lit.)

density

1.297 g/mL at 20 °C (lit.)

functional group

fluoro
hydroxyl
phenyl

SMILES string

OC(c1ccccc1)C(F)(F)F

InChI

1S/C8H7F3O/c9-8(10,11)7(12)6-4-2-1-3-5-6/h1-5,7,12H

InChI key

VNOMEAQPOMDWSR-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

183.2 °F - closed cup

Flash Point(C)

84 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service