779415
1-Azido-3-(trifluoromethyl)benzene solution
~0.5 M in tert-butyl methyl ether, ≥95.0% (HPLC)
Synonym(s):
3-(Trifluoromethyl)phenyl azide solution, m-Azido-α,α,α-trifluorotoluene solution
About This Item
Assay
≥95.0% (HPLC)
form
solution
concentration
~0.5 M in tert-butyl methyl ether
impurities
≤2.0% water
storage temp.
−20°C
SMILES string
FC(F)(F)c1cccc(c1)N=[N+]=[N-]
InChI
1S/C7H4F3N3/c8-7(9,10)5-2-1-3-6(4-5)12-13-11/h1-4H
InChI key
GIDLBMJGNCUXLD-UHFFFAOYSA-N
Application
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT RE 1
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
-18.4 °F
Flash Point(C)
-28 °C
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service