Skip to Content
Merck
All Photos(1)

Key Documents

750115

Sigma-Aldrich

2-Dicyclohexyl(2′,6′-dimethoxybiphenyl)phosphine gold(I) bis(trifluoromethylsulfonyl)imide

97%

Synonym(s):

SPhos AuNTf2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H35AuF6NO6PS2
CAS Number:
Molecular Weight:
887.64
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

Assay

97%

form

solid

reaction suitability

core: gold
reagent type: catalyst

mp

170-187 °C

SMILES string

FC(F)(F)S(=O)(=O)N([Au])S(=O)(=O)C(F)(F)F.COc1cccc(OC)c1-c2ccccc2P(C3CCCCC3)C4CCCCC4

InChI

1S/C26H35O2P.C2F6NO4S2.Au/c1-27-23-17-11-18-24(28-2)26(23)22-16-9-10-19-25(22)29(20-12-5-3-6-13-20)21-14-7-4-8-15-21;3-1(4,5)14(10,11)9-15(12,13)2(6,7)8;/h9-11,16-21H,3-8,12-15H2,1-2H3;;/q;-1;+1

InChI key

DCWMCSDFBBTUCM-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service