Skip to Content
Merck
All Photos(1)

Key Documents

741930

Sigma-Aldrich

Poly(N-isopropylacrylamide-co-acrylic acid)

acrylic acid 15 mol %

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H11NO)m (C3H4O2)n
UNSPSC Code:
12162002
NACRES:
NA.23

form

powder or crystals

composition

acrylic acid, 15 mol %

viscosity

7500-12500, 5 % in H2O

transition temp

LCST 32-36 °C (UVVI, water, pH 3-5)

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shunsuke Nakajima et al.
Soft matter, 13(20), 3710-3719 (2017-04-25)
Stimuli-responsive microfibers are fabricated by extruding mixed solutions of poly(N-isopropylacrylamide-co-acrylic acid) (pNIPAM-AAc) and sodium alginate (Na-alginate) using a microfluidic spinning system. The fabricated microfibers shrink and swell with temperature and/or pH. By controlling the extruded laminar flow, microfibers capable of

Articles

By altering the physicochemical properties, smart or intelligent drug delivery systems can be designed to deliver therapeutic molecules on-demand. Learn more about the application of stimuli-responsive materials in drug delivery.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service