Skip to Content
Merck
All Photos(1)

Documents

721867

Sigma-Aldrich

3-Bromo-1-methylpyrrolidine

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H10BrN
Molecular Weight:
164.04
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.491

bp

153-159 °C

density

1.361 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

CN1CCC(Br)C1

InChI

1S/C5H10BrN/c1-7-3-2-5(6)4-7/h5H,2-4H2,1H3

InChI key

ZHUYTHOKJZSZQJ-UHFFFAOYSA-N

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

129.9 °F

Flash Point(C)

54.4 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service