Skip to Content
Merck
All Photos(1)

Key Documents

700770

Sigma-Aldrich

(R)-SIPHOS-PE

97%

Synonym(s):

(11aR)-(+)-10,11,12,13-Tetrahydrodiindeno[7,1-de:1′, 7′-fg][1,3,2]dioxaphosphocin-5-bis[(R)-1-phenylethyl]amine, N-Di[(R)-1-phenylethyl]-[(R)-1,1′-spirobiindane-7,7′-diyl]-phosphoramidite

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C33H32NO2P
CAS Number:
Molecular Weight:
505.59
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

Assay

97%

form

powder

optical activity

[α]22/D +337°, c = 1 in chloroform

mp

90-96 °C

storage temp.

−20°C

InChI

1S/C33H32NO2P/c1-23(25-11-5-3-6-12-25)34(24(2)26-13-7-4-8-14-26)37-35-29-17-9-15-27-19-21-33(31(27)29)22-20-28-16-10-18-30(36-37)32(28)33/h3-18,23-24H,19-22H2,1-2H3/t23-,24-,33?/m1/s1

InChI key

ZXLQLIDJAURBDD-HRPAVAKOSA-N

Application

(R)-SIPHOS-PE can be used as:
  • A catalyst in the preparation of tricyclic alkaloid (+)-aphanorphine via asymmetric carboamination and Friedel−Crafts reaction.[1]
  • A ligand in the synthesis of chiral ketones via hydroacylation of olefins using salicylaldehyde derivatives.[2]
  • A ligand in the enantioselective synthesis of substituted pyrrolidine derivatives via Pd-catalyzed alkene aminoarylation reactions.[3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Recent developments in palladium-catalyzed alkene aminoarylation reactions for the synthesis of nitrogen heterocycles
Schultz DM and Wolfe JP
Synthesis, 44(03), 351-361 (2012)
Regio-and enantioselective intermolecular hydroacylation: Substrate-directed addition of salicylaldehydes to homoallylic sulfides
Coulter MM, et al.
Journal of the American Chemical Society, 132(46), 16330-16333 (2010)
Enantioconvergent synthesis of (+)-aphanorphine via asymmetric Pd-catalyzed alkene carboamination
Mai DN, et al.
Organic Letters, 13(11), 2932-2935 (2011)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service