Skip to Content
Merck
All Photos(1)

Key Documents

692875

Sigma-Aldrich

Aluminum nickel cobalt

alloy 5, disc, size 13 mm × 6 mm

Synonym(s):

AlNiCo

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352311
PubChem Substance ID:
NACRES:
NA.23

form

disc

composition

Aluminum, 7.5-8.7 wt. %
Cobalt, 23.5-25 wt. %
Copper, 2-3.5 wt. %
Iron, 45.3-52.9 wt. %
Nickel, 12.5-15.5 wt. %
Silicon, 1.5 wt. %
Titanium, 0.15-0.5 wt. %

size

13 mm × 6 mm

SMILES string

[Al].[Co].[Ni]

InChI

1S/Al.Co.Ni

InChI key

VKLQGCFJXKUNRB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Magnetic parameters/properties:
• Maximum Energy Product (BH): 40 kJ/m3 (5.0 MGsOe)
• Residual Induction (Br): 1.25 T (12.5 kGs)
• Coercive Force (Hc): 48 kA/m (0.60 kOe)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Magnetic materials find diverse applications from data storage to renewable energy.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service