Skip to Content
Merck
All Photos(2)

Key Documents

686662

Sigma-Aldrich

Polyester-16-hydroxyl-1-carboxyl bis-MPA dendron, generation 4

Synonym(s):

Dendron-G4-Carboxyl-OH

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12162002
NACRES:
NA.23

description

hydroxyl surface groups

form

powder

mol wt

generation 4

feature

focal point carboxylic acid

no. Surface Groups

16

storage temp.

2-8°C

General description

calculated mol. wt. 1,760

Other Notes

Bis-MPA = 2,2-Bis(hydroxyl-methyl)propionic acid

Legal Information

Manufactured by Polymer Factory Sweden AB.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Dendrons in nanomaterials - Find features and benefits of polyester dendrons and hyperbranched polymer products.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service