Skip to Content
Merck
All Photos(2)

Key Documents

680532

Sigma-Aldrich

3-(N,N-Dimethylamino)phenylboronic acid

≥95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H12BNO2
CAS Number:
Molecular Weight:
165.00
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95%

mp

178-190 °C

functional group

amine

SMILES string

CN(C)c1cccc(c1)B(O)O

InChI

1S/C8H12BNO2/c1-10(2)8-5-3-4-7(6-8)9(11)12/h3-6,11-12H,1-2H3

InChI key

YZQQHZXHCXAJAV-UHFFFAOYSA-N

General description

May contain varying amounts of anhydride

Application

Reactant involved in synthesis of different protein effector including:
  • Modulators of survival motor neuron protein
  • Glucokinase activators
  • Aryl ethers for use as Bacillus anthracis enoyl-ACP reductase inhibitors

Reactant involved in synthesis of:
  • Thiourea-functionalized paracyclophanes
  • Low-background fluorescent imaging agents for nitric oxide

Reactant to undergo regioselective iodination and bromination

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gözde Murat Saltan et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 188, 372-381 (2017-08-02)
In an approach to develop efficient organic optoelectronic devices to be used in light-driven systems, a series of three thiophene linked benzimidazole conjugates were synthesized and characterized. The combination of two thiophene rings to a benzimidazole core decorated with different

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service