Skip to Content
Merck
All Photos(1)

Key Documents

680109

Sigma-Aldrich

5-Methyl-2-furanboronic acid pinacol ester

95%

Synonym(s):

4,4,5,5-Tetramethyl-2-(5-methyl-furan-2-yl)-(1,3,2)dioxaborolane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H17BO3
CAS Number:
Molecular Weight:
208.06
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

refractive index

n20/D 1.476

density

1.023 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

Cc1ccc(o1)B2OC(C)(C)C(C)(C)O2

InChI

1S/C11H17BO3/c1-8-6-7-9(13-8)12-14-10(2,3)11(4,5)15-12/h6-7H,1-5H3

InChI key

FNPZFZKLYGWKLH-UHFFFAOYSA-N

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service