Skip to Content
Merck
All Photos(1)

Key Documents

670065

Sigma-Aldrich

Triethylmethylphosphonium dibutyl phosphate

≥97.0% (CH)

Synonym(s):

Methyltriethylphosphonium dibutyl phosphate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H36O4P2
CAS Number:
Molecular Weight:
342.39
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

≥97.0% (CH)

impurities

≤1.0% water

anion traces

bromide (Br-): ≤50 mg/kg
chloride (Cl-): ≤10 mg/kg
fluoride (F-): ≤10 mg/kg
nitrate (NO3-): ≤25 mg/kg
phosphate (PO43-): ≤25 mg/kg
sulfate (SO42-): ≤10 mg/kg

SMILES string

CC[P+](C)(CC)CC.CCCCOP([O-])(=O)OCCCC

InChI

1S/C8H19O4P.C7H18P/c1-3-5-7-11-13(9,10)12-8-6-4-2;1-5-8(4,6-2)7-3/h3-8H2,1-2H3,(H,9,10);5-7H2,1-4H3/q;+1/p-1

InChI key

KFJORSGSCYXKQU-UHFFFAOYSA-M

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service