Skip to Content
Merck
All Photos(3)

Key Documents

646164

Sigma-Aldrich

1-(4-Fluorobenzyl)piperazine

97%

Synonym(s):

1-[(4-Fluorophenyl)methyl]piperazine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H15FN2
CAS Number:
Molecular Weight:
194.25
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
form:
solid
Assay:
97%

Assay

97%

form

solid

functional group

fluoro

SMILES string

Fc1ccc(CN2CCNCC2)cc1

InChI

1S/C11H15FN2/c12-11-3-1-10(2-4-11)9-14-7-5-13-6-8-14/h1-4,13H,5-9H2

InChI key

OOSZCNKVJAVHJI-UHFFFAOYSA-N

Application

Precursor involved in the synthesis of a variety of biologically active molecules including:
  • Chemokine antagonists
  • Pyrido[1,2-a]benzimidazoles with cytotoxic and antiplasmodial activity
  • Pyrimidine derivatives as cholinesterase and Aβ-aggregation inhibitors
  • Chlorokojic acid derivatives with antibacterial and antiviral activities
  • EGFR and Aurora A kinase inhibitors
  • Neurokinin-2 receptor antagonists

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service