Skip to Content
Merck
All Photos(1)

Key Documents

609552

Sigma-Aldrich

Ethanolamine-15N

98 atom % 15N

Synonym(s):

2-Aminoethanol-15N

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCH2CH215NH2
CAS Number:
Molecular Weight:
62.08
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:

Note: This product can be packaged on demand. For information on pricing, availability and packaging of custom sizes, please contact Stable Isotopes Customer Service

isotopic purity

98 atom % 15N

Assay

99% (CP)

bp

170 °C (lit.)

mp

11 °C (lit.)

density

1.028 g/mL at 25 °C

mass shift

M+1

SMILES string

[15NH2]CCO

InChI

1S/C2H7NO/c3-1-2-4/h4H,1-3H2/i3+1

InChI key

HZAXFHJVJLSVMW-LBPDFUHNSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

199.4 °F

Flash Point(C)

93 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service