Skip to Content
Merck
All Photos(1)

Key Documents

587176

Sigma-Aldrich

Lanthanum(III) phosphate hydrate

≥99.99% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
LaPO4 · xH2O
CAS Number:
Molecular Weight:
233.88 (anhydrous basis)
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Assay

≥99.99% trace metals basis

form

chunks

reaction suitability

reagent type: catalyst
core: lanthanum

impurities

≤100 ppm Trace Rare Earth Analysis

SMILES string

[La+3].[H]O[H].[O-]P([O-])([O-])=O

InChI

1S/La.H3O4P.H2O/c;1-5(2,3)4;/h;(H3,1,2,3,4);1H2/q+3;;/p-3

InChI key

MXLPYXTVXABWIU-UHFFFAOYSA-K

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shigehiro Kagaya et al.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry, 19(7), 1061-1064 (2003-07-26)
Cadmium ranging from 1 - 8 ng could be coprecipitated quantitatively with lanthanum phosphate at pH 5 - 6 from up to 200 mL of river water samples spiked with 5 microg of indium as an internal standard. Cadmium and
Jian Feng et al.
Journal of magnetic resonance (San Diego, Calif. : 1997), 209(2), 300-305 (2011-02-26)
A new approach combining a long pulse with the DEPTH sequence (Cory and Ritchey, Journal of Magnetic Resonance, 1988) greatly improves the efficiency for suppressing probe background signals arising from spinning modules. By applying a long initial excitation pulse in
Biofunctionalization of fluorescent rare-earth-doped lanthanum phosphate colloidal nanoparticles.
Felix Meiser et al.
Angewandte Chemie (International ed. in English), 43(44), 5954-5957 (2004-11-18)
Shigehiro Kagaya et al.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry, 18(8), 923-926 (2002-08-31)
By applying an internal standardization, we could use a rapid coprecipitation technique using lanthanum phosphate as a coprecipitant for preconcentration of iron(III) and lead in their flame atomic absorption spectrometric determination. Indium as an internal standard was added to the
S T Gammon et al.
Analytical biochemistry, 369(1), 80-86 (2007-07-31)
Current methodologies for quantifying radiolabeled nucleoside monophosphates and nucleoside analogues result in high retention of unphosphorylated guanosine nucleosides in the case of lanthanum chloride precipitation or inconsistent retention of nucleotides in the case of DEAE cellulose filter papers. This study

Articles

Lanthanide ions in spectral conversion enhance solar cell efficiency via photon conversion.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service