Skip to Content
Merck
All Photos(2)

Key Documents

575518

Sigma-Aldrich

3-(4-Methoxyphenoxy)-1-propanesulfonyl chloride

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H3COC6H4O(CH2)3SO2Cl
CAS Number:
Molecular Weight:
264.73
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

45-46.7 °C (lit.)

SMILES string

COc1ccc(OCCCS(Cl)(=O)=O)cc1

InChI

1S/C10H13ClO4S/c1-14-9-3-5-10(6-4-9)15-7-2-8-16(11,12)13/h3-6H,2,7-8H2,1H3

InChI key

UFZBQBXKJZWBBV-UHFFFAOYSA-N

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service