Skip to Content
Merck
All Photos(1)

Documents

565784

Sigma-Aldrich

3-Bromo-5-fluoro-2-propoxyphenylboronic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H11BBrFO3
CAS Number:
Molecular Weight:
276.90
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

form

solid

mp

51-56 °C (lit.)

SMILES string

CCCOc1c(Br)cc(F)cc1B(O)O

InChI

1S/C9H11BBrFO3/c1-2-3-15-9-7(10(13)14)4-6(12)5-8(9)11/h4-5,13-14H,2-3H2,1H3

InChI key

XOBLNJQYFCQQTQ-UHFFFAOYSA-N

Other Notes

Contains varying amounts of anhydride

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eike-Benjamin Braune et al.
Breast cancer research : BCR, 26(1), 4-4 (2024-01-04)
Dysregulated Notch signalling contributes to breast cancer development and progression, but validated tools to measure the level of Notch signalling in breast cancer subtypes and in response to systemic therapy are largely lacking. A transcriptomic signature of Notch signalling would
Vivek Kumar et al.
Cells, 11(21) (2022-11-12)
Photon-based radiotherapy upregulates Notch signaling in cancer, leading to the acquisition of the stem cell phenotype and induction of invasion/migration, which contributes to the development of resistance to therapy. However, the effect of carbon ion radiotherapy (CIRT) on Notch signaling
Sandeep Gupta et al.
Cell reports, 40(3), 111119-111119 (2022-07-21)
Restoring sensation after injury or disease requires a reproducible method for generating large quantities of bona fide somatosensory interneurons. Toward this goal, we assess the mechanisms by which dorsal spinal interneurons (dIs; dI1-dI6) can be derived from mouse embryonic stem
Shota Shimizu et al.
Scientific reports, 11(1), 5199-5199 (2021-03-06)
The class II α-isoform of phosphatidylinositol 3-kinase (PI3K-C2α) plays a crucial role in angiogenesis at least in part through participating in endocytosis and, thereby, endosomal signaling of several cell surface receptors including VEGF receptor-2 and TGFβ receptor in vascular endothelial
Lai Wang et al.
Arthritis research & therapy, 25(1), 63-63 (2023-04-16)
We aimed to explore activation of the Notch signaling pathway in knee-innervating lumbar dorsal root ganglia (DRG) in the course of experimental osteoarthritis (OA) in mice, and its role in knee hyperalgesia. Cultured DRG cells were stimulated with the TLR4

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service