Skip to Content
Merck
All Photos(1)

Key Documents

55788

Sigma-Aldrich

Nα-Fmoc-Nε-Dabcyl-L-lysine

technical, ≥90% (HPLC)

Synonym(s):

Nε-4-[4-(Dimethylamino)phenylazo)benzoyl]-Nα-Fmoc-L-lysine, Fmoc-Lys(Dabcyl)-OH

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H37N5O5
CAS Number:
Molecular Weight:
619.71
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

grade

technical

Assay

≥90% (HPLC)

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

2-8°C

SMILES string

CN(C)c1ccc(cc1)\N=N\c2ccc(cc2)C(=O)NCCCC[C@H](NC(=O)OCC3c4ccccc4-c5ccccc35)C(O)=O

InChI

1S/C36H37N5O5/c1-41(2)27-20-18-26(19-21-27)40-39-25-16-14-24(15-17-25)34(42)37-22-8-7-13-33(35(43)44)38-36(45)46-23-32-30-11-5-3-9-28(30)29-10-4-6-12-31(29)32/h3-6,9-12,14-21,32-33H,7-8,13,22-23H2,1-2H3,(H,37,42)(H,38,45)(H,43,44)/b40-39+/t33-/m0/s1

InChI key

FPOPWTDBGMLRNG-JEFPPREJSA-N

Other Notes

Building block for the synthesis of fluorogenic protease substrates[1][2]

related product

Product No.
Description
Pricing

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Q.M. Wang et al.
Antiviral Chem. Chemother., 8, 303-303 (1997)
Automated Synthesis of Fluorogenic Protease Substrates: Design of Probes for Alzheimers Disease-Associated Proteases.
Wang, G.T., et al.
Bioorganic & Medicinal Chemistry Letters, 2, 1665-1665 (1992)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service