557668
2,2,4-Trimethyl-2H-imidazole 1-oxide
≥98%
Synonym(s):
TMIO
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C6H10N2O
CAS Number:
Molecular Weight:
126.16
Beilstein:
5424342
MDL number:
UNSPSC Code:
12171500
Assay
≥98%
mp
32-36 °C (lit.)
SMILES string
CC1=NC(C)(C)[N+]([O-])=C1
InChI
1S/C6H10N2O/c1-5-4-8(9)6(2,3)7-5/h4H,1-3H3
InChI key
CNIGTNPXWSLRPH-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Nitrone spin trapping agent of O-,C- and S-centered radicals.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service