Skip to Content
Merck
All Photos(1)

Key Documents

552232

Sigma-Aldrich

2,5-Dimethoxybenzenesulfonyl chloride

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3O)2C6H3SO2Cl
CAS Number:
Molecular Weight:
236.67
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

mp

112-116 °C (lit.)

SMILES string

COc1ccc(OC)c(c1)S(Cl)(=O)=O

InChI

1S/C8H9ClO4S/c1-12-6-3-4-7(13-2)8(5-6)14(9,10)11/h3-5H,1-2H3

InChI key

SHELADVIRCCTFN-UHFFFAOYSA-N

General description

2,5-Dimethoxybenzenesulfonyl chloride can be prepared from 2,5-dimethoxybenzenesulfonic acid.1 It can also be obtained from the reaction between chlorosulfonic acid and 1,4-dimethoxybenzene.2

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Chemistry of Thioether-Substituted Hydroquinones and Quinones. III. An Unexpected Rearrangement of a Heterocyclic Group.
Chapman DD, et al.
The Journal of Organic Chemistry, 30(5), 1520-1523 (1965)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service