Skip to Content
Merck
All Photos(2)

Documents

550698

Sigma-Aldrich

2,6-Diisopropyl-N,N-dimethylaniline

>90%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)2CH]2C6H3N(CH3)2
CAS Number:
Molecular Weight:
205.34
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

>90%

refractive index

n20/D 1.5 (lit.)

bp

86 °C/2.5 mmHg (lit.)

density

0.873 g/mL at 25 °C (lit.)

SMILES string

CC(C)c1cccc(C(C)C)c1N(C)C

InChI

1S/C14H23N/c1-10(2)12-8-7-9-13(11(3)4)14(12)15(5)6/h7-11H,1-6H3

InChI key

ALXIOUGHHXXLKX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2,6-Diisopropyl-N,N-dimethylaniline (DIDMA) is a tertiary amine. It can undergo iron-catalyzed oxidative amidation with propionaldehyde (PrCHO) to form the corresponding hindered amide.

Application

2,6-Diisopropyl-N,N-dimethylaniline may be used as a co-initiator in two-photon polymerization.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Two-photon polymerization of a diacrylate using fluorene photoinitiators-sensitizers.
Fitilis I, et al.
Journal of Photochemistry and Photobiology A: Chemistry, 215(1), 25-30 (2010)
One-and two-photon induced polymerization of Methylmethacrylate using colloidal CdS semiconductor quantum dots.
Strandwitz NC, et al.
Journal of the American Chemical Society, 130(26), 8280-8288 (2008)
Iron-Catalyzed Oxidative Amidation of Tertiary Amines with Aldehydes.
Li Y, et al.
Chemistry (Weinheim An Der Bergstrasse, Germany), 19(1), 82-86 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service