Skip to Content
Merck
All Photos(1)

Key Documents

547794

Sigma-Aldrich

Tris[N,N-bis(trimethylsilyl)amide]neodymium(III)

97%

Synonym(s):

1,1,1-Trimethyl-N-(trimethylsilyl)silanamine neodymium(III) salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Nd(N(Si(CH3)3)2)3
CAS Number:
Molecular Weight:
625.39
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

Assay

97%

form

solid

reaction suitability

reagent type: catalyst
core: neodymium

mp

156-159 °C (lit.)

SMILES string

C[Si](C)(C)N([Nd](N([Si](C)(C)C)[Si](C)(C)C)N([Si](C)(C)C)[Si](C)(C)C)[Si](C)(C)C

InChI

1S/3C6H18NSi2.Nd/c3*1-8(2,3)7-9(4,5)6;/h3*1-6H3;/q3*-1;+3

InChI key

LWKIKGQVWYVALF-UHFFFAOYSA-N

Application

Reagent for synthesis of:
  • Imidophosphinate complexes of lanthanides
  • Neodymium methoxides containing thienyl substituents
  • Phenylene-bridged binuclear organolanthanide complexes
  • Near-infrared photoluminescent neodymium(III) complexes

Reagent for:
  • Rare-earth metal carboxylate-based catalysts
  • Regioselective C-H activation

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1 - Skin Corr. 1B - Water-react 2

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

36.0 °F - closed cup

Flash Point(C)

2.2 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service