Skip to Content
Merck
All Photos(1)

Documents

546348

Sigma-Aldrich

2-(4-Bromophenyl)-6-(4-chlorophenyl)pyridine-4-carboxylic acid

97%

Synonym(s):

2-(4-Bromophenyl)-6-(4-chlorophenyl)isonicotinic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H11BrClNO2
CAS Number:
Molecular Weight:
388.64
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

291-294 °C (lit.)

SMILES string

OC(=O)c1cc(nc(c1)-c2ccc(Br)cc2)-c3ccc(Cl)cc3

InChI

1S/C18H11BrClNO2/c19-14-5-1-11(2-6-14)16-9-13(18(22)23)10-17(21-16)12-3-7-15(20)8-4-12/h1-10H,(H,22,23)

InChI key

APLDOGUHCNVFLZ-UHFFFAOYSA-N

General description

2-(4-Bromophenyl)-6-(4-chlorophenyl)pyridine-4-carboxylic acid [BPCPPCA] bears three functional groups: bromophenyl, chlorophenyl and carboxylic acid group. These groups facilitate the deposition of BPCPPCA molecules on the surface of a bulk insulator (calcite) at room temperature leading to hierarchical polymerization.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sequential and site-specific on-surface synthesis on a bulk insulator.
Kittelmann, Markus, et al.
ACS Nano, 7(6), 5614-5620 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service