Skip to Content
Merck
All Photos(1)

Key Documents

545589

Sigma-Aldrich

3-(Acetylthio)propionitrile

97%

Synonym(s):

3-Acetylsulfanylpropionitrile

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COS(CH2)2CN
CAS Number:
Molecular Weight:
129.18
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

refractive index

n20/D 1.4930 (lit.)

bp

246-247 °C (lit.)

density

1.120 g/mL at 25 °C (lit.)

SMILES string

CC(=O)SCCC#N

InChI

1S/C5H7NOS/c1-5(7)8-4-2-3-6/h2,4H2,1H3

InChI key

BMBATCYWJILFCR-UHFFFAOYSA-N

Application

3-(Acetylthio)propionitrile may be used in the synthesis of ethyl S-acetyl 3-mercaptopropionthioimidate ester hydrochloride.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F Dosio et al.
Bioconjugate chemistry, 9(3), 372-381 (1998-05-20)
Several immunotoxins (ITs) were synthesized by the attachment of clavin, a recombinant toxic protein derived from Aspergillus clavatus, to the monoclonal antibody Mgr6 that recognizes an epitope of the gp185(HER-2) extracellular domain expressed on breast and ovarian carcinoma cells. Conjugation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service