Skip to Content
Merck
All Photos(1)

Key Documents

522031

Sigma-Aldrich

Dichloromethane-d2

99.9 atom % D

Synonym(s):

Dideuteromethylenechloride, Methylene chloride-d2

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CD2Cl2
CAS Number:
Molecular Weight:
86.94
Beilstein:
1733318
EC Number:
MDL number:
UNSPSC Code:
12142201
PubChem Substance ID:

isotopic purity

99.9 atom % D

Quality Level

packaging

pk of 10 × 0.6 mL

refractive index

n20/D 1.422 (lit.)

bp

40 °C (lit.)

density

1.362 g/mL at 25 °C (lit.)

mass shift

M+2

SMILES string

[2H]C([2H])(Cl)Cl

InChI

1S/CH2Cl2/c2-1-3/h1H2/i1D2

InChI key

YMWUJEATGCHHMB-DICFDUPASA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service