499919
3-Formyl-2-thienylboronic acid
≥95%
Synonym(s):
3-Formyl-2-thiopheneboronic acid
About This Item
Recommended Products
Assay
≥95%
mp
330 °C (dec.) (lit.)
SMILES string
OB(O)c1sccc1C=O
InChI
1S/C5H5BO3S/c7-3-4-1-2-10-5(4)6(8)9/h1-3,8-9H
InChI key
HYXMHAHVUFTVFZ-UHFFFAOYSA-N
Application
- Suzuki-Miyaura cross-coupling reactions
Other Notes
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Suzuki-Miyaura cross-coupling reaction is extensively used in organic chemistry, polymer science, and pharmaceutical industries.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service