Skip to Content
Merck
All Photos(1)

Key Documents

491101

Sigma-Aldrich

1,3-Propanediol-2-13C

99 atom % 13C

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCH213CH2CH2OH
CAS Number:
Molecular Weight:
77.09
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

Note: This product can be packaged on demand. For information on pricing, availability and packaging of custom sizes, please contact Stable Isotopes Customer Service

isotopic purity

99 atom % 13C

Quality Level

refractive index

n20/D 1.44 (lit.)

bp

214 °C (lit.)

mp

−27 °C (lit.)

density

1.067 g/mL at 25 °C

mass shift

M+1

SMILES string

OC[13CH2]CO

InChI

1S/C3H8O2/c4-2-1-3-5/h4-5H,1-3H2/i1+1

InChI key

YPFDHNVEDLHUCE-OUBTZVSYSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service