About This Item
Assay
98%
refractive index
n20/D 1.588 (lit.)
bp
101 °C/0.1 mmHg (lit.)
density
1.168 g/mL at 25 °C (lit.)
SMILES string
O=C=Nc1ccccc1Oc2ccccc2
InChI
1S/C13H9NO2/c15-10-14-12-8-4-5-9-13(12)16-11-6-2-1-3-7-11/h1-9H
InChI key
KCHQIPPONLLUGU-UHFFFAOYSA-N
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 3
Flash Point(F)
235.4 °F - closed cup
Flash Point(C)
113 °C - closed cup
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service