Skip to Content
Merck
All Photos(1)

Key Documents

483281

Sigma-Aldrich

Chromium(III) bromide hexahydrate

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CrBr3 · 6H2O
CAS Number:
Molecular Weight:
399.80
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

Assay

99%

form

powder

mp

79 °C (lit.)

density

5.4 g/mL at 25 °C (lit.)

SMILES string

O.O.O.O.O.O.Br[Cr](Br)Br

InChI

1S/3BrH.Cr.6H2O/h3*1H;;6*1H2/q;;;+3;;;;;;/p-3

InChI key

XIOAYKRRSJTLOZ-UHFFFAOYSA-K

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Repr. 2 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service