Skip to Content
Merck
All Photos(1)

Key Documents

476463

Sigma-Aldrich

Isopropyl nitrite

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H7NO2
CAS Number:
Molecular Weight:
89.09
EC Number:
MDL number:
UNSPSC Code:
12352103

Assay

95%

SMILES string

CC(C)ON=O

Pictograms

FlameSkull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-20.2 °F - closed cup

Flash Point(C)

-29 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service