Skip to Content
Merck
All Photos(1)

Key Documents

468177

Sigma-Aldrich

[3-((((Heptadecafluorooctyl)sulfonyl)amino)propy)]trimethylammonium iodide solution

42 wt. % in isopropanol/water

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3(CF2)7SO2NH(CH2)3N(CH3)3I
CAS Number:
Molecular Weight:
726.23
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
Pricing and availability is not currently available.

contains

17% water

concentration

42 wt. % in isopropanol/water

refractive index

n20/D 1.39

pH

3-5

viscosity

30 cP(25 °C)(lit.)

bp

82 °C

density

1.2 g/mL at 25 °C

SMILES string

[I-].C[N+](C)(C)CCCNS(=O)(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F

Application

Cationic fluorosurfactant.
Leveling agent for polishes and coatings.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service