Skip to Content
Merck
All Photos(2)

Key Documents

437921

Sigma-Aldrich

4,4′-(9-Fluorenylidene)bis(2-phenoxyethanol)

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H26O4
CAS Number:
Molecular Weight:
438.51
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

117-120 °C (lit.)

SMILES string

OCCOc1ccc(cc1)C3(c2ccc(OCCO)cc2)c4ccccc4-c5ccccc35

InChI

1S/C29H26O4/c30-17-19-32-23-13-9-21(10-14-23)29(22-11-15-24(16-12-22)33-20-18-31)27-7-3-1-5-25(27)26-6-2-4-8-28(26)29/h1-16,30-31H,17-20H2

InChI key

NQXNYVAALXGLQT-UHFFFAOYSA-N

General description

4,4′-(9-Fluorenylidene)bis(2-phenoxyethanol) is an aryldiol.[1][2]

Application

4,4′-(9-Fluorenylidene)bis(2-phenoxyethanol) may be used for the following syntheses:
  • poly-(carbotetramethyldisiloxane)s, containing poly[oxy{4,4′-(9-fluorenylidene)bis(2-phenoxyethylene)}oxytetramethyldisiloxane] (a fluorescent aromatic chromophore group)[1]
  • poly[oxy(arylene)oxy(tetramethyldisilylene)]s, containing fluorescent aromatic chromophore groups in the polymer main chain, via melt copolymerization reaction with 1,2-bis(diethylamino)tetramethyldisilane[2]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Melt Copolymerization Reactions between 1, 3-Bis (diethylamino) tetramethyldisiloxane and Aryldiol Derivatives.
Jung IK and Park YT.
Bull. Korean Chem. Soc., 32(4), 1303-1309 (2011)
Synthesis and Properties of Poly [oxy (arylene) oxy (tetramethyldisilylene)] s via Melt Copolymerization Reaction.
Jung EA and Park YT.
Bull. Korean Chem. Soc., 34(6), 1637-1642 (2013)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service