Skip to Content
Merck
All Photos(2)

Key Documents

430811

Sigma-Aldrich

Polylauryllactam-block-polytetrahydrofuran

melt index 7-11 g/10 min (235°C/1kg)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[NH(CH2)11CO]x(OCH2CH2CH2CH2)yOH
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

form

pellets

melt index

7-11 g/10 min (235°C/1kg)

hardness

25 (Shore D)

mp

148 °C

transition temp

softening point 60 °C (Vicat, ASTM D 1525)

density

1.01 g/mL at 25 °C (lit.)

SMILES string

OCCCCO.O=C1CCCCCCCCCCCN1

Looking for similar products? Visit Product Comparison Guide

Application

Flexible sporting goods, hoses and semi-conductors (with fillers).

Features and Benefits

Thermoplastic elastomer. Undergoes repeated flexure with limited deformation. High abrasion resistance.

Physical form

Alternating block copolymer.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Macromolecules, 26, 6350-6353 (1993)
Hatfield, G.R. et al.
Macromolecules, 26, 6350-6353 (1993)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service