Skip to Content
Merck
All Photos(1)

Key Documents

374954

Sigma-Aldrich

Diethylene glycol monododecyl ether

97%

Synonym(s):

2-(2-Dodecyloxyethoxy)ethanol, C12E2, Dodecyldiglycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)11(OCH2CH2)2OH
CAS Number:
Molecular Weight:
274.44
Beilstein:
1765704
EC Number:
MDL number:
UNSPSC Code:
12352103
Pricing and availability is not currently available.

Assay

97%

refractive index

n20/D 1.448 (lit.)

bp

168-169 °C/0.8 mmHg (lit.)

mp

165 °C (lit.)

density

0.904 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCCCCCOCCOCCO

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service