Skip to Content
Merck
All Photos(2)

Key Documents

367990

Sigma-Aldrich

1H-Benzotriazol-1-ylmethyl isocyanide

94%

Synonym(s):

1-(Isocyanomethyl)-1H-benzotriazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H6N4
CAS Number:
Molecular Weight:
158.16
Beilstein:
7762310
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22
Assay:
94%

Assay

94%

reaction suitability

reaction type: C-C Bond Formation

mp

80-84 °C (lit.)

functional group

amine
isonitrile

SMILES string

[C-]#[N+]Cn1nnc2ccccc12

InChI

1S/C8H6N4/c1-9-6-12-8-5-3-2-4-7(8)10-11-12/h2-5H,6H2

InChI key

YZTNZXMSOPEFKC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant for:
  • Passerini condensation
  • Ugi type reactions
  • Heterocyclization with imines
  • Preparation of silver (isocyanomethyl)benzotriazole polymeric complex

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service