Skip to Content
Merck
All Photos(1)

Key Documents

339636

Sigma-Aldrich

exo-N-Hydroxy-7-oxabicyclo[2.2.1]hept-5-ene-2,3-dicarboximide

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H7NO4
CAS Number:
Molecular Weight:
181.15
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

98%

mp

194 °C (dec.) (lit.)

solubility

THF: soluble 10 mg/mL, clear, colorless

SMILES string

ON1C(=O)[C@H]2C3OC(C=C3)[C@H]2C1=O

InChI

1S/C8H7NO4/c10-7-5-3-1-2-4(13-3)6(5)8(11)9(7)12/h1-6,12H/t3-,4+,5?,6?

InChI key

PPVGNPSAUJFBJY-LAXKNYFCSA-N

General description

exo-N-Hydroxy-7-oxabicyclo[2.2.1]hept-5-ene-2,3-dicarboximide undergoes cross-coupling with aryl boronic acid for ROMP-based O-alkylhydroxylamine parallel synthesis.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jamerson Carneiro de Oliveira et al.
Carbohydrate polymers, 250, 116966-116966 (2020-10-15)
The Diels-Alder reaction is a promising click chemistry for the design of advanced materials from cellulose nanocrystals (CNCs). Transferring such chemistry to cellulose nanocrystals requires the precise grafting of reactive Diels-Alder moeities under heterogeneous conditions without compromising the nanocrystals morphology.
Florence S Gaucher-Wieczorek et al.
Journal of combinatorial chemistry, 12(5), 655-658 (2010-09-14)
The parallel synthesis of O-aryloxyamines remains an unfulfilled need in the field of medicinal chemistry and fragment-based approaches. To fill this gap a solution-phase two-step process based on (1) a copper-catalyzed cross-coupling of aryl boronic acids with a fluorous tagged

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service