Skip to Content
Merck
All Photos(2)

Key Documents

335568

Sigma-Aldrich

2-Methoxy-2-phenylethanol

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH(OCH3)CH2OH
CAS Number:
Molecular Weight:
152.19
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

liquid

refractive index

n20/D 1.519 (lit.)

bp

237 °C (lit.)

density

1.061 g/mL at 25 °C (lit.)

functional group

ether
hydroxyl
phenyl

SMILES string

COC(CO)c1ccccc1

InChI

1S/C9H12O2/c1-11-9(7-10)8-5-3-2-4-6-8/h2-6,9-10H,7H2,1H3

InChI key

JDTUPLBMGDDPJS-UHFFFAOYSA-N

Application

(S)-2-methoxy-2-phenylethanol was employed as chiral probe for the determination of absolute configuration of active (-) enantiomer of amlodipine.[1] 2-Methoxy-2-phenylethanol was also used in the synthesis of optically active 1,4-dihydropyridines.[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

208.4 °F - closed cup

Flash Point(C)

98 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enzymatic resolution of the chiral inductor 2-methoxy-2-phenylethanol.
Monterde MI, et al.
Tetrahedron Asymmetry, 13(10), 1091-1096 (2002)
S Goldmann et al.
Journal of medicinal chemistry, 35(18), 3341-3344 (1992-09-04)
The active (-) enantiomer of amlodipine was originally reported to have R configuration. This does not concur with other 1,4-dihydropyridines with known absolute configuration. This configuration has now been determined by X-ray structural analysis using (1S)-camphanic acid and (S)-2-methoxy-2-phenylethanol as

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service