Skip to Content
Merck
All Photos(1)

Key Documents

332976

Sigma-Aldrich

4-Propylbenzoic acid

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2C6H4CO2H
CAS Number:
Molecular Weight:
164.20
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

142-144 °C (lit.)

functional group

carboxylic acid

SMILES string

CCCc1ccc(cc1)C(O)=O

InChI

1S/C10H12O2/c1-2-3-8-4-6-9(7-5-8)10(11)12/h4-7H,2-3H2,1H3,(H,11,12)

InChI key

ATZHGRNFEFVDDJ-UHFFFAOYSA-N

Application

4-Propylbenzoic acid was used in the preparation of [2,7-dimethoxy-8-(4-propylbenzoyl)naphthalen-1-yl](4-propylphenyl)-methanone.[1]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kosuke Sasagawa et al.
Acta crystallographica. Section E, Structure reports online, 68(Pt 8), o2596-o2596 (2012-08-21)
In the title compound, C(32)H(32)O(4), the 4-propyl-benzoyl groups at the 1- and 8-positions of the naphthalene ring system are aligned almost anti-parallel, and their benzene rings make a dihedral angle of 8.64 (10)°. The dihedral angles between the naphthalene ring system

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service