Skip to Content
Merck
All Photos(1)

Documents

283738

Sigma-Aldrich

Potassium aquapentachlororuthenate(III)

Synonym(s):

Dipotassium aquapentachlororuthenate(III)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
K2RuCl5(H2O)
CAS Number:
Molecular Weight:
374.55
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

form

powder

Quality Level

concentration

26-30% Ru (gravimetric)

technique(s)

H-NMR: suitable

SMILES string

O.[K+].[K+].Cl[Ru--](Cl)(Cl)(Cl)Cl

InChI

1S/5ClH.2K.H2O.Ru/h5*1H;;;1H2;/q;;;;;2*+1;;+3/p-5

InChI key

SRVGBBZEPHUDFZ-UHFFFAOYSA-I

Looking for similar products? Visit Product Comparison Guide

Application

The product may be used for eletrodeposition of bifunctional catalyst films, PtRu on nanoporous TiO2 substrates.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nanoporous TiO 2-supported bimetallic catalysts for methanol oxidation in acidic media.
Hepel M, et al.
Electrochemical Communications, 8(9), 1439-1444 (2006)
Sunder N Dhuri et al.
Dalton transactions (Cambridge, England : 2003), 44(16), 7634-7642 (2015-03-27)
A mononuclear high-valent trans-dioxoruthenium(VI) complex, trans-[Ru(VI)(TMC)(O)2](2+) (TMC = 1,4,8,11-tetramethyl-1,4,8,11-tetraazacyclotetradecane), was synthesized and characterized by various spectroscopic techniques and X-ray crystallography. The reactivity of the trans-[Ru(VI)(TMC)(O)2](2+) complex was investigated in hydride transfer and hydrogen atom transfer reactions. The mechanism of hydride

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service