All Photos(1)
About This Item
Linear Formula:
CH3(CH2)3CH=CHCH3
CAS Number:
Molecular Weight:
98.19
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Recommended Products
vapor pressure
87 mmHg ( 37.7 °C)
Assay
97%
form
liquid
refractive index
n20/D 1.407 (lit.)
bp
98-99 °C (lit.)
density
0.708 g/mL at 25 °C (lit.)
SMILES string
[H]\C(C)=C(/[H])CCCC
InChI
1S/C7H14/c1-3-5-7-6-4-2/h3,5H,4,6-7H2,1-2H3/b5-3-
InChI key
OTTZHAVKAVGASB-HYXAFXHYSA-N
General description
The rate constants for the epoxidation of cis-2-heptene with meta-chloroperbenzoic acid (mCPBA) and [2-percarboxyethyl]-functionalized silica in different solvents were studied.
Application
cis-2-Heptene (alkene) was used in the synthesis of the corresponding bis(10-phenoxathiiniumyl)alkane adduct.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
21.2 °F - closed cup
Flash Point(C)
-6 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Rossella Mello et al.
Organic & biomolecular chemistry, 12(20), 3246-3250 (2014-04-15)
The rate constants for the epoxidation of cis-2-heptene with [2-percarboxyethyl]-functionalized silica (1a) and meta-chloroperbenzoic acid (mCPBA) (1b) in different solvents have been determined at temperatures in the -10 to 40 °C range. The heterogeneous epoxidation exhibits a dependence of the
Bing-Jun Zhao et al.
The Journal of organic chemistry, 72(16), 6154-6161 (2007-07-03)
Addition of phenoxathiin cation radical (PO*+) to acyclic alkenes in acetonitrile (MeCN) solution occurred stereospecifically to form bis(10-phenoxathiiniumyl)alkane adducts. Stereospecific trans addition is ascribed to the intermediacy of an episulfonium cation radical. The alkenes used were cis- and trans-2-butene, cis-
Hiroyuki Ohno et al.
Journal of AOAC International, 97(5), 1452-1458 (2015-04-24)
A headspace GC/MS method was developed for identification and quantitation of residual volatile organic compounds in poly(methyl methacrylate) (PMMA) kitchen utensils. A sample was cut into small pieces, then N,N-dimethylacetamide was added in a headspace vial and sealed. After storing
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service