Skip to Content
Merck
All Photos(1)

Documents

244856

Sigma-Aldrich

Potassium

ingot, 99.95% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
K
CAS Number:
Molecular Weight:
39.10
EC Number:
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:
NACRES:
NA.23

vapor pressure

0.09 mmHg ( 260 °C)

Assay

99.95% trace metals basis

form

ingot

reaction suitability

reagent type: reductant

resistivity

6.1 μΩ-cm, 20°C

bp

760 °C (lit.)

mp

64 °C (lit.)

density

0.86 g/mL at 25 °C (lit.)

SMILES string

[K]

InChI

1S/K

InChI key

ZLMJMSJWJFRBEC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Combines under pressure with silver metal to form 2 new compounds: K2Ag and K3Ag.†

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B - Water-react 1

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Glucose-insulin-potassium revived: current status in acute coronary syndromes and the energy-depleted heart.
Alexandra N Grossman et al.
Circulation, 127(9), 1040-1048 (2013-03-06)
Pieter Clauw et al.
Plant physiology, 167(3), 800-816 (2015-01-22)
Although the response of plants exposed to severe drought stress has been studied extensively, little is known about how plants adapt their growth under mild drought stress conditions. Here, we analyzed the leaf and rosette growth response of six Arabidopsis
Jun Ueda et al.
Molecular and cellular biology, 34(19), 3702-3720 (2014-07-30)
Hypoxia promotes stem cell maintenance and tumor progression, but it remains unclear how it regulates long-term adaptation toward these processes. We reveal a striking downregulation of the hypoxia-inducible histone H3 lysine 9 (H3K9) demethylase JMJD1A as a hallmark of clinical
Jordan E Bisanz et al.
Applied and environmental microbiology, 81(15), 4965-4975 (2015-05-17)
The nutritional status of pregnant women is vital for healthy outcomes and is a concern for a large proportion of the world's population. The role of the microbiota in pregnancy and nutrition is a promising new area of study with
Stefan D Anker et al.
European journal of heart failure, 17(10), 1050-1056 (2015-05-27)
Hyperkalaemia in heart failure patients limits use of cardioprotective renin-angiotensin-aldosterone system inhibitors (RAASi). Sodium zirconium cyclosilicate (ZS-9) is a selective potassium ion trap, whose mechanism of action may allow for potassium binding in the upper gastrointestinal tract as early as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service