Skip to Content
Merck
All Photos(1)

Key Documents

228540

Sigma-Aldrich

(Dimethylamino)acetonitrile

97%

Synonym(s):

N,N-Dimethylglycinonitrile

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2NCH2CN
CAS Number:
Molecular Weight:
84.12
Beilstein:
1735677
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
form:
liquid
Assay:
97%

vapor pressure

760 mmHg ( 137 °C)

Assay

97%

form

liquid

refractive index

n20/D 1.410 (lit.)

bp

137-138 °C (lit.)

density

0.863 g/mL at 25 °C (lit.)

SMILES string

CN(C)CC#N

InChI

1S/C4H8N2/c1-6(2)4-3-5/h4H2,1-2H3

InChI key

PLXBWEPPAAQASG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

(Dimethylamino)acetonitrile (N,N-Dimethylglycinonitrile) was used in the synthesis of α-dimethylamino-β-oxonitriles.

Pictograms

FlameSkull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Oral - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

95.0 °F - closed cup

Flash Point(C)

35 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aroylations of N, N-dimethylglycinonitrile to form. alpha.-dimethylamino-. beta.-oxonitriles. Cyclizations with acetophenone to give 3-dimethylamino-2-pyridones.
The Journal of Organic Chemistry, 33(6), 2562-2564 (1968)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service