227684
Potassium hexacyanoferrate(II) trihydrate
99%, ACS reagent
Synonym(s):
Potassium ferrocyanide, Yellow prussiate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
grade
ACS reagent
Assay
98.5-102.0%
99%
impurities
≤0.005% insolubles
mp
70 °C (lit.)
anion traces
chloride (Cl-): ≤0.01%
sulfate (SO42-): passes test
SMILES string
O.O.O.[K+].[K+].[K+].[K+].N#C[Fe-4](C#N)(C#N)(C#N)(C#N)C#N
InChI
1S/6CN.Fe.4K.3H2O/c6*1-2;;;;;;;;/h;;;;;;;;;;;3*1H2/q;;;;;;-4;4*+1;;;
InChI key
NRYOVBQTVIWDNU-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Autophagy, 11(10), 1891-1904 (2015-08-28)
Metastatic renal cell carcinomas (mRCC) are highly vascularized tumors that are a paradigm for the treatment with antiangiogenesis drugs targeting the vascular endothelial growth factor (VEGF) pathway. The available drugs increase the time to progression but are not curative and
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service