Skip to Content
Merck
All Photos(1)

Key Documents

220825

Sigma-Aldrich

Propyl 1-(1-phenylethyl)imidazole-5-carboxylate hydrochloride

99%

Synonym(s):

Propoxate hydrochloride, R 7464

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H18N2O2 · HCl
CAS Number:
Molecular Weight:
294.78
MDL number:

Assay

99%

SMILES string

Cl.CCCOC(=O)c1cncn1C(C)c2ccccc2

Application

Potent fish anesthetic, about 100 times as potent as tricaine methanesulfonate (cat. no. E10521).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nature, 205, 1018-1018 (1965)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service