Skip to Content
Merck
All Photos(1)

Key Documents

207233

Supelco

Lead atomic absorption standard solution

1,000 μg/mL Pb in 1 wt. % HNO3 (Exact Pb concentration on label)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
PubChem Substance ID:

concentration

1,000 μg/mL Pb in 1 wt. % HNO3 (Exact Pb concentration on label)

technique(s)

AAS: suitable

density

1.01 g/mL at 25 °C

format

single component solution


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service