Skip to Content
Merck
All Photos(2)

Documents

149969

Sigma-Aldrich

Ethyl 3-(1-adamantyl)-3-oxopropionate

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H22O3
CAS Number:
Molecular Weight:
250.33
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.499 (lit.)

bp

108-110 °C/0.06 mmHg (lit.)

density

1.037 g/mL at 25 °C (lit.)

SMILES string

CCOC(=O)CC(=O)C12C[C@H]3C[C@H](C[C@H](C3)C1)C2

InChI

1S/C15H22O3/c1-2-18-14(17)6-13(16)15-7-10-3-11(8-15)5-12(4-10)9-15/h10-12H,2-9H2,1H3/t10-,11+,12-,15-

InChI key

FOISHGXCIBGQJU-WUQLGEGHSA-N

Application

Ethyl 3-(1-adamantyl)-3-oxopropionate may be used in chemical synthesis studies.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service